missing translation for 'onlineSavingsMsg'
Learn More

Titanium Thinfoil, 0.0114mm (0.00045in) thick, 99.6% (metals basis)

Catalog No. p-7059240
Change view
Click to view available options
Quantity:
100 x 100 mm
200 x 200 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44242GH 100 x 100 mm
AA44242GW 200 x 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA44242GH Supplier Thermo Scientific Chemicals Supplier No. 044242GH
Only null left

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 100 x 100 mm
Odor Odorless
Weight ≈0.514g/100x100mm
Form Thin Foil
Percent Purity 99.6%
Grade Commercial Pure 4
Assay (metals basis)
Chemical Name or Material Titanium Thinfoil, 0.0114mm (0.00045 in.) thick, Light Tight
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only