missing translation for 'onlineSavingsMsg'
Learn More

Titanium sponge, 3mm (0.12in) and down, 99.5% (metals basis)

Catalog No. p-7052059
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4245930 250 g
AA42459A1 1 kg
AA42459A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4245930 Supplier Thermo Scientific Chemicals Supplier No. 04245930
Only null left

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 250 g
Odor Odorless
Particle Size 3mm (0.12 in.) & down
Form Sponge
UN Number UN2878
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Titanium sponge
Hazard Category Warning
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: TITANIUM SPONGE GRANULES
EINECSNumber 231-142-3
RTECSNumber XR1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only