missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium slug, 9.5mm (0.37in) dia x 9.5mm (0.37in) length, 99.995% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Supplier: Thermo Scientific Chemicals 041303KJ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Titanium slug | |
| 7440-32-6 | |
| Odorless | |
| Slug | |
| Ti | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| [Ti] | |
| 47.87 | |
| CHEBI:33341 |
| 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length | |
| 5 Pc. | |
| ≈3g/slug | |
| (metals basis) | |
| MFCD00011264,MFCD00151301 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| titanium | |
| 23963 | |
| 99.995% |
Chemical Identifiers
| 7440-32-6 | |
| 47.87 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| 23963 | |
| titanium |
| Ti | |
| MFCD00011264,MFCD00151301 | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| CHEBI:33341 | |
| [Ti] |
Safety and Handling
EINECSNumber : 231-142-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only