missing translation for 'onlineSavingsMsg'
Learn More

Titanium rod, 6.4mm (0.25in) dia, 99.99% (metals basis)

Catalog No. p-4782201
Change view
Click to view available options
Quantity:
5 cm
25 cm
4 x 25 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14001BQ 5 cm
AA14001BU 25 cm
AA14001CZ 4 x 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA14001BQ Supplier Thermo Scientific Chemicals Supplier No. 014001BQ
Only null left

Form: Approx. 1.45g/cm | CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Silver to Gray
Appearance Silver Gray rod
Density 4.506g/cm3
Quantity 5 cm
Odor Odorless
Form Rod
Assay Percent Range 99.99% (Metals basis)
Chemical Name or Material Titanium
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only