missing translation for 'onlineSavingsMsg'
Learn More

Titanium rod, 12.7mm (0.5in) dia, 99.99% (metals basis)

Catalog No. p-7049322
Change view
Click to view available options
Quantity:
10 cm
2 cm
Catalog No. Quantity
AA13999BP 2 cm
AA13999BS 10 cm
2 options

Catalog No. AA13999BP

Supplier: Thermo Scientific Chemicals 013999BP

Only null left

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 2 cm
Odor Odorless
Weight ≈5.7g/cm
Form Rod
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Titanium rod, 12.7mm (0.5 in.) dia.
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only