Learn More
Titanium powder, spherical, -150 mesh, 99.9% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Supplier: Thermo Scientific Chemicals 04154522
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Titanium powder | |
| 100 g | |
| Odorless | |
| (metals basis) | |
| MFCD00011264,MFCD00151301 | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| [Ti] | |
| 47.87 | |
| CHEBI:33341 |
| 7440-32-6 | |
| −150 Mesh | |
| Spherical Powder | |
| Ti | |
| UN2546 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| titanium | |
| 23963 | |
| 99.9% |
Chemical Identifiers
| 7440-32-6 | |
| 47.87 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| 23963 | |
| titanium |
| Ti | |
| MFCD00011264,MFCD00151301 | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| CHEBI:33341 | |
| [Ti] |
Safety and Handling
GHS H Statement
H251
Self-heating: may catch fire.
P210-P222-P280-P335+P334-P370+P378q
H250-H500
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: TITANIUM POWDER, DRY
EINECSNumber : 231-142-3
RTECSNumber : XR1700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only