missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, dehydrided, -325 mesh, 99.99% (metals basis)

Catalog No. p-4824993
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3667914 25 g
AA3667922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3667914 Supplier Thermo Scientific Chemicals Supplier No. 03667914
Only null left

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Gray
Appearance Dark Gray powder
Quantity 25 g
Mesh Size -325 Mesh
Odor Odorless
Form Dehydrided Powder
Assay Percent Range 99.99% (Metals basis)
Chemical Name or Material Titanium
Hazard Category H250-H500
Precautionary Statement P210-P222-P280-P335+P334-P370+P378q
DOTInformation DOT Class: 4.2, Packaging Group: II
EINECSNumber 231-142-3
RTECSNumber XR1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only