missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, -325 mesh, 99% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol

Supplier:  Thermo Scientific Chemicals 01038622

 View more versions of this product

Catalog No. AA1038622


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gray
7440-32-6
100 g
Odorless
99% (Metals basis)
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
Titanium
Dark Gray powder
-325 Mesh
Powder
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
47.87
CHEBI:33341

Chemical Identifiers

7440-32-6
47.87
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264,MFCD00151301
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P222-P280-P335+P334-P370+P378q

H250-H500

DOTInformation : DOT Class: 4.2, Packaging Group: II

EINECSNumber : 231-142-3

RTECSNumber : XR1700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only