missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

$86.86 - $308.93

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4310218
View Documents
Thermo Scientific Chemicals
04310218
50 g
Each for $86.86
Only null left
 
AA4310230
View Documents
Thermo Scientific Chemicals
04310230
250 g
Each for $308.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-32-6
47.87
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264,MFCD00151301
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]

Specifications

7440-32-6
−200 Mesh
Powder
MFCD00011264,MFCD00151301
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
47.87
CHEBI:33341
Titanium powder
50 g
Odorless
Ti
UN2546
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
99.5%
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H251
Self-heating: may catch fire.

P210-P222-P280-P335+P334-P370+P378q

H250-H500

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: TITANIUM POWDER, DRY

EINECSNumber : 231-142-3

RTECSNumber : XR1700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only