missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) Oxide, Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. p-4939069
Change view
Click to view available options
Quantity:
1 kg
25 kg
50 kg
Packaging:
Fibre carton
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1008081000 1 kg Plastic bottle
M1008089025 25 kg Fibre carton
M1008089050 50 kg Fibre carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1008081000 Supplier Technidata Supplier No. 1.00808.1000
Only null left
For analysis

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Physical Form Powder
Quantity 1 kg
Formula Weight 79.87
Grade Reagent Ph. Eur.
Odor Odorless
Packaging Plastic bottle
Appearance White powder
Chemical Name or Material Titanium(IV) oxide