Learn More
Titanium(IV) oxide, high surface area, 99.9% (metals basis)
CAS: 13463-67-7 | O2Ti | 79.87 g/mol
$183.59 - $718.48
Chemical Identifiers
| CAS | 13463-67-7 |
|---|---|
| Molecular Formula | O2Ti |
| Molecular Weight (g/mol) | 79.87 |
| MDL Number | MFCD00011269 MFCD00210650 |
| InChI Key | GWEVSGVZZGPLCZ-UHFFFAOYSA-N |
| Synonym | titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance |
| PubChem CID | 26042 |
| ChEBI | CHEBI:32234 |
| IUPAC Name | dioxotitanium |
| SMILES | O=[Ti]=O |
Description
Photocatalyst, catalyst, catalyst supportTitanium(IV) oxide is used in paints, varnishes, paper, plastics printing inks, sun screen, fibers, rubber, cosmetic products, anti-fogging coatings, foodstuffs, pure titanium, glass, glass ceramics, memristor, electrical ceramics, catalysts, electric conductors and chemical intermediates. It exhibit excellent photocatalytic properties (Honda-Fujishima effect) In addition, titanium dioxide is also used to cleave protein at the site where proline is present.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13463-67-7 | |
| 79.87 | |
| GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
| 26042 | |
| dioxotitanium |
| O2Ti | |
| MFCD00011269 MFCD00210650 | |
| titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
| CHEBI:32234 | |
| O=[Ti]=O |
Specifications
| 2500°C to 3000°C | |
| 13463-67-7 | |
| Powder | |
| 99.9% metal basis | |
| MFCD00011269 MFCD00210650 | |
| Insoluble in water,HCl,HNO3,dilute H2SO4; soluble in hot,conc. H2SO4,HF,and alkaliesSlightly soluble in hydrofluoric acid and hot concentrated sulfuric acid. Insoluble in water,hydrochloric acid,dilute sulfuric acid,and organic solvents. | |
| O=[Ti]=O | |
| 79.87 | |
| CHEBI:32234 | |
| Odorless | |
| Titanium(IV) oxide, High surface Area: S.A. 180-300 m2/g |
| 1830°C to 1850°C | |
| White | |
| 2 g | |
| O2Ti | |
| titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
| GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
| dioxotitanium | |
| 26042 | |
| 79.9 | |
| 4.26 |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
EINECSNumber : 236-675-5
RTECSNumber : XR2275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only