Learn More
Titanium(IV) oxide bis(2,4-pentanedionate)
CAS: 14024-64-7 | C10H16O5Ti | 264.1 g/mol
$140.34 - $2962.18
Chemical Identifiers
| CAS | 14024-64-7 |
|---|---|
| Molecular Formula | C10H16O5Ti |
| Molecular Weight (g/mol) | 264.1 |
| MDL Number | MFCD00013505 |
| InChI Key | ADVORQMAWLEPOI-UHFFFAOYSA-N |
| Synonym | bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide |
| PubChem CID | 74765597 |
| IUPAC Name | 4-hydroxypent-3-en-2-one;oxotitanium |
| SMILES | CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA7713009
|
Thermo Scientific Chemicals
07713009 |
10 g |
Each for $140.34
|
|
|||||
|
AA7713018
|
Thermo Scientific Chemicals
07713018 |
50 g |
Each for $616.10
|
|
|||||
|
AA7713030
|
Thermo Scientific Chemicals
07713030 |
250 g |
Each for $2,962.18
|
|
|||||
Description
Employed as a cross linking agent for cellulose fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14024-64-7 | |
| 264.1 | |
| ADVORQMAWLEPOI-UHFFFAOYSA-N | |
| 74765597 | |
| CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti] |
| C10H16O5Ti | |
| MFCD00013505 | |
| bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide | |
| 4-hydroxypent-3-en-2-one;oxotitanium |
Specifications
| 184°C (decomposition) | |
| Powder | |
| C10H16O5Ti | |
| bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide | |
| ADVORQMAWLEPOI-UHFFFAOYSA-N | |
| 4-hydroxypent-3-en-2-one;oxotitanium | |
| 74765597 | |
| Titanium (IV) oxide bis (2,4-pentanedionate) |
| 14024-64-7 | |
| 10 g | |
| MFCD00013505 | |
| Soluble in water (6.6g/L (20°C)). | |
| CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti] | |
| 264.1 | |
| 262.12 |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 237-861-9
RTECSNumber : XR2330000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only