missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide, 99.999%, (trace metal basis), predominantly rutile, Thermo Scientific Chemicals

Catalog No. p-221754
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC194340050 5 g Glass bottle
AC194340250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC194340050 Supplier Thermo Scientific Chemicals Supplier No. 194340050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

Boiling Point 2900.0°C
Melting Point 1855.0°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.999%
Linear Formula TiO2
Merck Index 15,9628
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
Formula Weight 79.88
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.23
Density 4.2300g/mL
Chemical Name or Material Titanium(IV) oxide, 99.999%
Show More Show Less
EINECSNumber 236-675-5
RTECSNumber XR2275000
TSCA TSCA

RUO – Research Use Only