missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(IV) oxide, 98+%, anatase powder
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 2900.0°C |
| Melting Point | 1855.0°C |
| Color | White |
| pH | 7.3 to 8.3 (aq. suspension) |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 98% min |
| Linear Formula | TiO2 |
| Merck Index | 15, 9628 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf |
| Show More |
Safety and Handling
| EINECSNumber | 236-675-5 |
| RTECSNumber | XR2275000 |
| TSCA | TSCA |
RUO – Research Use Only