missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 99+%

Catalog No. p-7080342
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA7712409 10 g
AA7712422 100 g
AA7712436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA7712409 Supplier Thermo Scientific Chemicals Supplier No. 07712409
Only null left

CAS: 5593-70-4 | C16H40O4Ti | 344.359 g/mol

Titanium(IV) n-butoxide is used in the preparation of nanosized titania powders in the anatase form and ferroelectric bismuth titanate thin films. It is also used as intermediates, paint additives, coating additives, processing aids and as process regulators. It is involved in the preparation of nanocrystalline titanium dioxide powders at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5593-70-4
Molecular Formula C16H40O4Ti
Molecular Weight (g/mol) 344.359
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Specifications

Boiling Point 310°C to 314°C
Melting Point -55°C
Color Yellow
Physical Form Liquid
Quantity 10 g
Linear Formula Ti[O(CH2)3CH3]4
UN Number UN2924
Beilstein 4148236
Flash Point 41°C (170°F)
Solubility Information Miscible with aliphatic,aromatic,chlorinated and oxygenated solvents.
Formula Weight 340.36
Percent Purity ≥99%
Odor Alcohol-like
Refractive Index 1.49
Sensitivity Moisture sensitive
Density 0.998 g/mL
Chemical Name or Material Titanium (IV) n-butoxide
Show More Show Less
Hazard Category H226-H315-H318-H335-H336
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 227-006-8
RTECSNumber XR1585000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only