missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) n-butoxide, 98+%

Catalog No. p-200627
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2 kg
10 kg
Packaging:
Glass bottle
Plastic Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC223192500 250 g Glass bottle
AC223195000 500 g Glass bottle
AC223190010 1 kg Glass bottle
AC223190020 2 kg Glass bottle
AC223190100 10 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC223192500 Supplier Thermo Scientific Chemicals Supplier No. 223192500

CAS: 5593-70-4 | C16H36O4Ti | 340.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 5593-70-4
Molecular Formula C16H36O4Ti
Molecular Weight (g/mol) 340.35
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Specifications

Melting Point -75°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 310.0°C to 314.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.486
Linear Formula Ti[O(CH2)3CH3]4
Quantity 250 g
Specific Gravity 0.99
Solubility Information Solubility in water: rapidly hydrolized by water. Other solubilities: miscible with most common organic solvents
Viscosity 50-80 mPa.s (20°C)
Formula Weight 340.35
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Titanium(IV) n-butoxide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
EINECSNumber 227-006-8
RTECSNumber XR1585000
TSCA TSCA
Recommended Storage the product darkens with age

RUO – Research Use Only