missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) chloride, 1M solution in dichloromethane, AcroSeal™

Catalog No. p-3213250
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377741000 100 mL AcroSeal™ Glass bottle
AC377748000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC377741000 Supplier Thermo Scientific Chemicals Supplier No. 377741000
Only null left

CAS: 7550-45-0 | Cl4Ti | 189.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Specifications

Chemical Name or Material Titanium(IV) chloride
Name Note 1M Solution in Dichloromethane
Color Brown to Yellow
CAS 7550-45-0, 75-09-2
Physical Form Solution
Quantity 100 mL
Molecular Formula Cl4Ti
MDL Number MFCD00011267
Merck Index 15, 9634
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
Solubility Information Solubility in water: reacts
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
IUPAC Name titanium(4+);tetrachloride
Molecular Weight (g/mol) 189.71
PubChem CID 160960
Formula Weight 189.71
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.365
Density 1.3650g/mL
Show More Show Less

RUO – Research Use Only