missing translation for 'onlineSavingsMsg'
Learn More

Titanium(III) sulfate, 20% in 1-4% sulfuric acid

Catalog No. AA39664A7
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39664A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39664A7 Supplier Thermo Scientific Chemicals Supplier No. 039664A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10343-61-0 | O12S3Ti2 | 383.902 g/mol

Titanium(III) sulfate is used in the manufacture of bulk, large scale chemicals (including petroleum products), fine chemicals, basic metals, including alloys, computer, electronic and optical products, electrical equipment. It is also used as a biochemical for proteomics research. It is a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10343-61-0
Molecular Formula O12S3Ti2
Molecular Weight (g/mol) 383.902
MDL Number MFCD00036317
InChI Key SOBXOQKKUVQETK-UHFFFAOYSA-H
Synonym titanium sesquisulfate, titanium iii sulfate, unii-69hci1s2xi, 69hci1s2xi, dititanium tris sulphate, titanium 3+ sulfate 2:3, titan iii-sulfat, acmc-1bolh, sulfuric acid, titanium 3+ salt 3:2
PubChem CID 165866
IUPAC Name titanium(3+);trisulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ti+3].[Ti+3]

Specifications

Physical Form Liquid
Quantity 5 kg
UN Number UN3264
Merck Index 14,9476
Solubility Information Fully miscible with water.
Formula Weight 383.38
Concentration or Composition (by Analyte or Components) 20% in 1 to 4% sulfuric acid
Density 1.15 g/mL
Chemical Name or Material Titanium(III) sulfate
Hazard Category H315-H319-H350
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only