Learn More
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5%
Used as a catalyst in the production of polypropylene. | CAS: 12003-13-3 | AlCl12Ti3 | 595.98 g/mol
$241.85 - $1057.42
Chemical Identifiers
| CAS | 12003-13-3 |
|---|---|
| Molecular Formula | AlCl12Ti3 |
| Molecular Weight (g/mol) | 595.98 |
| MDL Number | MFCD00145412 |
| InChI Key | KSKPGAGLAZXNEO-UHFFFAOYSA-H |
| IUPAC Name | titanium(4+) aluminium(3+) heptachloride |
| SMILES | Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl |
Description
Titanium(III) chloride-Aluminum(III) chloride, TiCl{3} 76.0-78.5% is used as a catalyst in the production of polypropylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12003-13-3 | |
| 595.98 | |
| KSKPGAGLAZXNEO-UHFFFAOYSA-H | |
| Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl |
| AlCl12Ti3 | |
| MFCD00145412 | |
| titanium(4+) aluminium(3+) heptachloride |
Specifications
| 12003-13-3 | |
| 100 g | |
| AlCl12Ti3 | |
| Titanium aluminum chloride | |
| KSKPGAGLAZXNEO-UHFFFAOYSA-H | |
| titanium(4+) aluminium(3+) heptachloride | |
| 596.15 | |
| Titanium(III) Chloride-Aluminum(III) Chloride |
| Powder | |
| 76.0 to 78.5% | |
| MFCD00145412 | |
| Insoluble in water. | |
| Cl[Al](Cl)Cl.Cl[Ti](Cl)Cl | |
| 595.98 | |
| Air and moisture sensitive |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 234-421-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only