missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid
CAS: 7705-07-9 | Cl3Ti | 154.22 g/mol
Supplier: Thermo Scientific Chemicals 395620250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
| Titanium(III) chloride | |
| Purple | |
| 25 mL | |
| TiCl3 | |
| 15,9637 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether | |
| [Cl-].[Cl-].[Cl-].[Ti+3] | |
| 154.22 | |
| ∼20% (TiCl3) (exact strength stated on certificate of analysis) | |
| 1.2000g/mL |
| 7705-07-9,7647-01-0,7732-18-5 | |
| Liquid | |
| Cl3Ti | |
| MFCD00011266 | |
| Titanous chloride | |
| YONPGGFAJWQGJC-UHFFFAOYSA-K | |
| titanium(3+) trichloride | |
| 154.26 | |
| Plastic bottle |
RUO – Research Use Only