missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) oxide pellets, 99.9% (metals basis)

Catalog No. AA4294022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4294022 100 g
AA4294014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4294022 Supplier Thermo Scientific Chemicals Supplier No. 04294022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12137-20-1 | OTi | 63.87 g/mol

Used for vacuum deposition. Also applications include sunscreens and UV-blocking in cosmetics, varnishes, wood preservatives, textile fibers, and packaging films. Also used as paint pigment, opacifying agent, welding rod fluxes, optical coatings, as a catalyst, ceramic finish coat, plastics, elastomers, coated fabrics, printing inks, roofing granules, glass, and in glazes. Also identified as Titanium monoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12137-20-1
Molecular Formula OTi
Molecular Weight (g/mol) 63.87
MDL Number MFCD00036279
InChI Key OGIDPMRJRNCKJF-UHFFFAOYSA-N
Synonym titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
PubChem CID 61685
IUPAC Name oxotitanium
SMILES O=[Ti]

Specifications

Boiling Point >3000°C
Melting Point 1700°C
Quantity 100 g
Odor Odorless
Density 4.95 g/mL
Solubility Information Insoluble in water.
Formula Weight 63.9
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Titanium (II) oxide pellets, 8 to 9mm(0.32 to 0.36 in.) dia. x 6 to 7mm(0.24 to 0.28 in.) thick
EINECSNumber 235-236-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only