missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) oxide, 99.9% (metals basis)

Catalog No. p-4782207
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4293936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4293936 Supplier Thermo Scientific Chemicals Supplier No. 04293936
Only null left

CAS: 12137-20-1 | OTi | 63.87 g/mol

Titanium(II) oxide associated with vanadium oxide forms a transition-metal-oxide monolayer catalyst, which is used in the selective oxidation and ammoxidation of alkylaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12137-20-1
Molecular Formula OTi
Molecular Weight (g/mol) 63.87
MDL Number MFCD00036279
InChI Key OGIDPMRJRNCKJF-UHFFFAOYSA-N
Synonym titanium monoxide, titanium ii oxide, titanium oxide tio, oxido titanium, titanium ii oxide pellets, titanium oxide, 99.9 %, titanium ii oxide, optical grade, titanium ii oxide trace metals basis 5g, titanium ii oxide, coating quality, granular, titanium ii oxide, pieces trace metals basis
PubChem CID 61685
IUPAC Name oxotitanium
SMILES O=[Ti]

Specifications

Boiling Point >3000°C
Melting Point 1700°C
Physical Form Lump
Quantity 500 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in concentrated sulfuric acid,concentrated hydrofluoric acid and alkali. Insoluble in water,diluted sulfuric acid,hydrochloric acid,nitric acid and organic solvents.
Formula Weight 63.9
Density 4.95
Chemical Name or Material Titanium(II) Oxide
EINECSNumber 235-236-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only