missing translation for 'onlineSavingsMsg'
Learn More

Titanium(II) hydride, 99% (metals basis)

Widley used as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. | CAS: 7704-98-5 | H2Ti

Supplier:  Thermo Scientific Chemicals 01285722

 View more versions of this product

Catalog No. AA1285722


Only null left
Explore available promotions

Description

Description

Titanium(II) hydride is widely utilized as an additive in the manufacturing of Alnico sintered magnets and in sintered powdered metals. It is used in the production of metal foam, powdered titanium metal and in pyrotechnics. It is involved in various applications such as ceramics, pyrotechnics and sports equipment. It is also used as a laboratory reagent and as a blowing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Titanium(II) hydride
7704-98-5
100 g
TiH2
UN1871
49.92
Plastic bottle
3.9
450°C (decomposition)
Powder
H2Ti
MFCD00151301
Soluble in water.
99%
Air and moisture sensitive
3.9

Chemical Identifiers

7704-98-5
H2Ti
MFCD00151301
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 231-726-8

RTECSNumber : XR2130000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only