missing translation for 'onlineSavingsMsg'
Learn More

Titanium gauze, 20 mesh woven from 0.076mm (0.003in) dia wire

Catalog No. AA40918FL
Change view
Click to view available options
Quantity:
75 x 75 mm
300 x 300 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40918FL 75 x 75 mm
AA40918HB 300 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40918FL Supplier Thermo Scientific Chemicals Supplier No. 040918FL

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 75 x 75 mm
Mesh Size 20 Mesh
Odor Odorless
Form Wire Cloth
Chemical Name or Material Titanium gauze, Woven from 0.076mm (0.003 in.) dia. wire
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only