missing translation for 'onlineSavingsMsg'
Learn More

Titanium foil, 0.74mm (0.029in) thick, annealed, 99.4+% (metals basis)

Catalog No. p-7048181
Change view
Click to view available options
Quantity:
200 x 300 mm
300 x 1000 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00029GX 200 x 300 mm
AA00029ZP 300 x 1000 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA00029GX Supplier Thermo Scientific Chemicals Supplier No. 000029GX
Only null left

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Gray
Quantity 200 x 300 mm
Odor Odorless
Weight ≈200g/200x300mm
Form Foil
Percent Purity ≥99.4%
Assay (metals basis)
Chemical Name or Material Titanium foil, 0.74mm (0.029 in.) thick, Annealed
EINECSNumber 231-142-3
RTECSNumber XR1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only