missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium foil, 0.5mm (0.02in) thick, annealed, 99% (metals basis)
Form: approx. 22.5g/100x100mm | CAS: 7440-32-6 | Ti | 47.87 g/mol
$100.99 - $1113.90
Chemical Identifiers
| CAS | 7440-32-6 |
|---|---|
| Molecular Formula | Ti |
| Molecular Weight (g/mol) | 47.87 |
| MDL Number | MFCD00011264,MFCD00151301 |
| InChI Key | RTAQQCXQSZGOHL-UHFFFAOYSA-N |
| Synonym | oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 |
| PubChem CID | 23963 |
| ChEBI | CHEBI:33341 |
| IUPAC Name | titanium |
| SMILES | [Ti] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA43677GH
|
Thermo Scientific Chemicals
043677GH |
100 x 100 mm |
Each for $100.99
|
|
|||||
|
AA43677GW
|
Thermo Scientific Chemicals
043677GW |
200 x 200 mm |
Each for $263.03
|
|
|||||
|
AA43677ZB
|
Thermo Scientific Chemicals
043677ZB |
200 x 1000 mm |
Each for $1,113.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-32-6 | |
| 47.87 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| 23963 | |
| titanium |
| Ti | |
| MFCD00011264,MFCD00151301 | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| CHEBI:33341 | |
| [Ti] |
Specifications
| Silver to Gray | |
| Silver Gray foil | |
| Odorless | |
| Foil | |
| MFCD00011264,MFCD00151301 | |
| RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
| titanium | |
| 23963 | |
| Titanium, Annealed |
| 7440-32-6 | |
| 100 x 100 mm | |
| 99% (Metals basis) | |
| Ti | |
| oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
| [Ti] | |
| 47.87 | |
| CHEBI:33341 |
Safety and Handling
EINECSNumber : 231-142-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only