missing translation for 'onlineSavingsMsg'
Learn More

Titanium foil, 0.25mm (0.01in) thick, 99.99+% (metals basis)

Catalog No. p-4408011
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog No. Quantity
AA13975FF 25 x 25 mm
AA13975FI 50 x 50 mm
AA13975GH 100 x 100 mm
3 options

Catalog No. AA13975FF

Supplier: Thermo Scientific Chemicals 013975FF

Only null left

Form: approx. 0.7g/25x25mm | CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Silver to Gray
Appearance Silver Gray foil
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Assay Percent Range 99.99+% (Metals basis)
Chemical Name or Material Titanium
EINECSNumber 231-142-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only