missing translation for 'onlineSavingsMsg'
Learn More

Titanium foil, 0.127mm (0.005in) thick, annealed, 99% (metals basis)

CAS: 7440-32-6 | Ti | 47.87 g/mol

$89.67 - $273.62

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA00971FI
View Documents
Thermo Scientific Chemicals
000971FI
50 x 50 mm N/A
Only null left
 
AA00971GT
View Documents
Thermo Scientific Chemicals
000971GT
150 x 150 mm N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-32-6
47.87
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264,MFCD00151301
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]

Specifications

7440-32-6
99% (metals basis)
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
Titanium foil
50 x 50 mm
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
47.87
CHEBI:33341
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only