missing translation for 'onlineSavingsMsg'
Learn More

Titanium Dioxide, Technical, Spectrum™ Chemical

Catalog No. p-7158640
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607421 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607421 Supplier Spectrum Chemical Mfg Cor Supplier No. T10812.5KG
Only null left

T1081, 13463-67-7, TiO2

Spectrum™ Chemical Titanium Dioxide, Technical , is the naturally available oxide of titanium also known as titania and is used as a pigment in paints, food coloring, cosmetics and in the production of pure titanium, electrical ceramics, catalysts and other chemical applications.

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Grade Technical
Packaging Poly Bottle