missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium Dioxide, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 12 kg |
| Grade | Reagent |
| Loss on Ignition | 0.003 |
| Packaging | Poly Pail |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.