missing translation for 'onlineSavingsMsg'
Learn More

Titanium Dioxide, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7158388
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607416 500 g Poly Bottle
18607417 2.5 kg Poly Pail
18607418 12 kg Poly Pail
18607419 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18607416 Supplier Spectrum Chemical Mfg Cor Supplier No. T1083500GMPL
Only null left

T1083, 13463-67-7, TiO2

Spectrum™ Chemical Titanium Dioxide, FCC is used as a food coloring and has an E-number, E171. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13463-67-7
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.87
MDL Number MFCD00011269 MFCD00210650
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
IUPAC Name dioxotitanium
SMILES O=[Ti]=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.0 to 100.5%
Grade FCC
Loss on Ignition 0.005
Packaging Poly Bottle