missing translation for 'onlineSavingsMsg'
Learn More

Titanium boride, Thermo Scientific Chemicals

$99.58 - $849.11

Chemical Identifiers

CAS 12045-63-5
Molecular Formula B2Ti
Molecular Weight (g/mol) 69.487
MDL Number MFCD00049595
InChI Key QYEXBYZXHDUPRC-UHFFFAOYSA-N
Synonym titanium boride,
PubChem CID 131664302
IUPAC Name boron;titanium
SMILES [B].[B].[Ti]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1136418
View Documents
Thermo Scientific Chemicals
01136418
50 g
Each for $99.58
Only null left
 
AA1136430
View Documents
Thermo Scientific Chemicals
01136430
250 g
Each for $308.93
Only null left
 
AA11364A1
View Documents
Thermo Scientific Chemicals
011364A1
1 kg
Each for $849.11
Only null left
 
Description

Description

Titanium boride is used in seals wear parts and cutting tools. It is used to make ballistic armour due to its high hardness. It is used to constitute composite materials in which the presence of the material serves to increase strength and fracture toughness of the matrix.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12045-63-5
69.487
QYEXBYZXHDUPRC-UHFFFAOYSA-N
131664302
[B].[B].[Ti]
B2Ti
MFCD00049595
titanium boride,
boron;titanium

Specifications

3225°C
Powder
B2Ti
titanium boride,
QYEXBYZXHDUPRC-UHFFFAOYSA-N
boron;titanium
131664302
4.38
12045-63-5
50 g
MFCD00049595
Soluble in hydrogen peroxide and sulfuric acid.
[B].[B].[Ti]
69.487
69.5
Titanium Boride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-961-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only