missing translation for 'onlineSavingsMsg'
Learn More

Titanium boride, Thermo Scientific Chemicals

Catalog No. AA1136418
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1136418 50 g
AA1136430 250 g
AA11364A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1136418 Supplier Thermo Scientific Chemicals Supplier No. 01136418

Titanium boride is used in seals wear parts and cutting tools. It is used to make ballistic armour due to its high hardness. It is used to constitute composite materials in which the presence of the material serves to increase strength and fracture toughness of the matrix.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12045-63-5
Molecular Formula B2Ti
Molecular Weight (g/mol) 69.487
MDL Number MFCD00049595
InChI Key QYEXBYZXHDUPRC-UHFFFAOYSA-N
Synonym titanium boride,
PubChem CID 131664302
IUPAC Name boron;titanium
SMILES [B].[B].[Ti]

Specifications

Melting Point 3225°C
Physical Form Powder
Quantity 50 g
Solubility Information Soluble in hydrogen peroxide and sulfuric acid.
Formula Weight 69.5
Density 4.38
Chemical Name or Material Titanium Boride
EINECSNumber 234-961-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only