missing translation for 'onlineSavingsMsg'
Learn More

Titanium aluminide, 99.5% (metals basis)

Used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products. | CAS: 12004-78-3 | Al3Ti | 128.812 g/mol

Supplier:  Thermo Scientific Chemicals 02289522

 View more versions of this product

Catalog No. AA2289522


Only null left
Explore available promotions

Description

Description

Titanium aluminide is used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Titanium aluminide
Powder
Al3Ti
UN3178
Insoluble in water.
[Al].[Al].[Al].[Ti]
128.812
99.5%
12004-78-3
100 g
MFCD00054094
titanium aluminum, aluminum, compd. with titanium 3:1, aluminum; titanium, aluminum, compound with titanium 3:1
OQPDWFJSZHWILH-UHFFFAOYSA-N
aluminum;titanium
6336852
Plastic bottle

Chemical Identifiers

12004-78-3
128.812
OQPDWFJSZHWILH-UHFFFAOYSA-N
6336852
[Al].[Al].[Al].[Ti]
Al3Ti
MFCD00054094
titanium aluminum, aluminum, compd. with titanium 3:1, aluminum; titanium, aluminum, compound with titanium 3:1
aluminum;titanium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: II

EINECSNumber : 234-461-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only