missing translation for 'onlineSavingsMsg'
Learn More

Titanium aluminide, -325 mesh, 99.5% (metals basis)

Catalog No. p-7080457
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8839309 10 g
AA8839318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA8839309 Supplier Thermo Scientific Chemicals Supplier No. 08839309

CAS: 12003-96-2 | AlTi3+3 | 170.583 g/mol

It finds use in several applications including automobiles and aircraft. Also used in air frames, investment casting, turbocharger rotors, and wire/fiber metal matrix composites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12003-96-2
Molecular Formula AlTi3+3
Molecular Weight (g/mol) 170.583
MDL Number MFCD00210656
InChI Key JRDVYNLVMWVSFK-UHFFFAOYSA-N
Synonym aluminum 3+ ion trititanium, aluminum, compd. with titanium 1:1
PubChem CID 44153015
IUPAC Name aluminum;titanium
SMILES [Al+3].[Ti].[Ti].[Ti]

Specifications

Quantity 10 g
UN Number UN3089
Solubility Information Insoluble in water.
Percent Purity 99.5%
Odor Odorless
Sensitivity Air Sensitive
Assay (metals basis)
Chemical Name or Material Titanium aluminide
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only