missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titan Yellow, TCI America™
Specifications
Specifications
| Formula Weight | 695.71 |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Titan Yellow |
Safety and Handling
| EINECSNumber | (5)-4156 |
| RTECSNumber | DL6423000 |
| TSCA | Yes |