Learn More
Andwin Scientific Inc TISSUETEK 24 SLIDE HOLDER PK/6 DFS Item
Supplier:  Andwin Scientific Inc AND4465
Histological staining apparatus, Tissue-Tek, 24 Microscope Slide Holder with Handle, For staining histological and cytological specimens. 12 solution wells deep enough for immersion of 1x3 in. microscope slides. The set comes complete with a metal frame, a s
Catalog No. NC0260316
Only null left
Explore available promotions