Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes in Tube Packs
Slotted Cassettes are ideal for use with routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in twelve color varieties.
Supplier: Epredia™ B851720BL
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Chemical Identifiers
| Yes | |
| Slots | |
| Yes | |
| TissueLoc™ Slotted Cassette |
| 750/Cs. | |
| Separate Lid, Back Hinge | |
| For Use With PrintMate™ Printers |
Specifications
| Slots | |
| Yes | |
| Yes | |
| For Use With PrintMate™ Printers |
| 750/Cs. | |
| Separate Lid, Back Hinge | |
| Blue | |
| TissueLoc™ Slotted Cassette |