Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes
For routine tissue samples, cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Supplier: Epredia™ B851954TN
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
| Slots | |
| 1000/Cs. | |
| Separate Lid, Back Hinge | |
| Tan |
| Yes | |
| Loose Packed | |
| No | |
| Slotted Cassette |