Learn More
Andwin Scientific Inc TISSUE-TEK SLIDE STAIN SET DFS Item
Supplier:  Andwin Scientific Inc AND4451
The Tissue-Tek Slide Staining Set is ideal for use in histological or cytological staining. This product offers concurrent utilization of up to 12 solution wells. It also features a snap-locking cover on solvent-resistant metal frame that eliminates the risk of spills. This set contains 10 white polypropylene dishes, 2 green solvent-resistant dishes, 12 lids, 12-unit metal carrier, and drain tray/cover. It is sold individually.
Catalog No. 50-899-90328
Only null left
Explore available promotions