missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes

Catalog No. 361041651
Change view
Click to view available options
Color:
Blue
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
361041651 Blue
361041650 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361041651 Supplier Fisherbrand™ Supplier No. M5036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes are innovative, disposable and make biopsy pads obsolete.

  • Molded from a special high-density polymer
  • Mesh design features more than 2000, 0.26 mm ''pores'' for maximum fluid exchange and drainage
  • One piece, integral lid eliminates the need for separate lids
  • Lids can be opened or closed as many times as needed and relocked securely each time
  • 45° anterior writing surface suitable for use with certain types of labeling instruments
  • Six-Compartment Cassette holds up to six tissue specimens; each one placed in a separate 0.25 x 0.43 in. (7 x 12 mm) numbered compartment

Specifications

Pore Style Mesh
Writing Face Angle 45°
Color Blue
No. of Compartments 6
Material High Density Polymer
Product Type Biopsy Cassette