missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Tissue Path™ Microsette™ Single Compartment Biopsy Cassettes

Catalog No. p-92304
Change view
Click to view available options
Color:
Green
Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15182706B Pink
15182706C Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15182706B
Only null left

Tissue Path™ Microsette™ Single Compartment Biopsy Cassettes are innovative, disposable, and make biopsy pads obsolete.

  • Molded from a special high-density polymer
  • Mesh design features more than 2000, 0.26 mm ''pores'' for maximum fluid exchange and drainage
  • One piece, integral lid eliminates the need for separate lids
  • Lids can be opened or closed as many times as needed and relocked securely each time
  • 45° anterior writing surface suitable for use with certain types of labeling instruments
  • Single Compartment Cassette measures 25 x 30 mm

Specifications

Pore Style Mesh
Writing Face Angle 45°
Closure Type Integral Lid
Color Pink
No. of Compartments 1
Material High Density Polymer
Product Type Biopsy Cassette