Learn More
Fisherbrand™ Tissue Path™ II Cassettes
Standard lids fit securely onto cassettes.
Supplier:  Fisherbrand™ 58127
 View more versions of this product
Catalog No. 15-120-400D
Only null left
Description
Description
  • Molded from a special high-density polymer resistant to all commonly used histological solvents
  • Slanted writing surface accepts markings easily, permitting positive sample identification
Order Info
Packaged in three dispenser boxes of 500
Specifications
Specifications
Separate Lid
Blue
High Density Polymer
Tissue Cassette
Product Suggestions
Product Suggestions