Learn More
Fisherbrand™ Tissue Path™ II Cassettes
Description
- Molded from a special high-density polymer resistant to all commonly used histological solvents
- Slanted writing surface accepts markings easily, permitting positive sample identification
Order Info
Packaged in three dispenser boxes of 500
Specifications
Specifications
| Closure Type | Separate Lid |
| Color | Pink |
| Material | High Density Polymer |
| Product Type | Tissue Cassette |