missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Tissue Marking Dye

Catalog No. STLSL6628GR
Change view
Click to view available options
Color:
Black
Blue
Green
Orange
Red
Violet
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STLSL6628GR Green
STLSL6628BK Black
STLSL6628BL Blue
STLSL6628OR Orange
STLSL6628RD Red
STLSL6628VT Violet
STLSL6628YL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSL6628GR Supplier StatLab™ Supplier No. SL6628GR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or inhaled.

Permanent tissue marking dyes formulated to define margins without bleeding or fading.

  • Proprietary formulation designed to eliminate evaporation and sludgy dyes, with fewer drips and spills
  • Durable, bright dyes hold permanently through processing and staining procedures
  • Unique lids may either be completely removed or conveniently flipped open to prevent drying

Specifications

Color Green
Product Type Tissue Marking Dye
Quantity 8 oz. Bottle
Solution Type Mordant Dye
For Use With (Application) Tissue Marking
Form Liquid