missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ Tissue-Clearing Reagent TOMEI (for plants)

Catalog No. T3530100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3530100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3530100ML Supplier TCI America™ Supplier No. T3530100ML

missing translation for 'validMainframeMsg'

The transparency technique TOMEI for plant analysis was developed by Prof. Matsunaga et al. at Tokyo University of Science. This method enables the clearing of many plants in just a few hours and clear observation of deeper internal structures.

  • TOMEI allows plant specimen internal structures to be observed clearly without dissection
  • Simple and rapid clearing of plant can be performed following the basic procedure
  • Deeper structures can be observed by using confocal microscope
  • Clears many plant specimens in just hours

Specifications

For Use With (Equipment) Confocal Laser-scanning Microscopy (CLSM) Device
Product Line TOMEI
Quantity 100 mL
For Use With (Application) Plant Cytology and Histological research
Form Liquid