missing translation for 'onlineSavingsMsg'
Learn More

TCI America™ Tissue-Clearing Reagent iTOMEI-D

Catalog No. T39405ML
Change view
Click to view available options
Capacity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
T39405ML 5 mL
T394025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T39405ML Supplier TCI America™ Supplier No. T39405ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improved TOMEI (iTOMEI) is a method that specializes the TOMEI clearing process for fluorescent protein observation.

  • Enables transparency by simple osmosis in just a few days
  • Keeps the fluorescence of a fluorescent protein such as GFP or tdTomato co-expressed as a reporter gene
  • Suppresses auto-fluorescence
  • Applicable to wide variety of plant species including Oryza sativa, Arabidopsis thaliana, Marchantia polymorpha

Specifications

Capacity 5 mL
For Use With (Equipment) Confocal Laser-scanning Microscopy (CLSM) Device
Product Line TOMEI
For Use With (Application) Plant Cytology and Histological research
Form Liquid