missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-X1
CUBIC-X1 is an optional addition to the CUBIC clearing process, and can help swell/expand tissues for improved visibility. Used in combination with CUBIC-X2 (T3867).
$91.35 - $292.32
Specifications
| For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
|---|---|
| Product Line | CUBIC |
| For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
| Form | Liquid |
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|
T386625ML
|
TCI America™
T386625ML |
25 mL |
Each for $91.35
|
|
|||||
|
T3866100ML
|
TCI America™
T3866100ML |
100 mL |
Each for $292.32
|
|
|||||
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-X1 is an optional addition to the CUBIC clearing protocol, and can be used to swell/expand tissue for improved visibility.
Note: Both CUBIC-X1 (T3866) and CUBIC-X2 (T3867) are required for the CUBIC clearing process with expansion/swelling.
Specifications
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |