missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-R+(M)
Used along with CUBIC-L (T3740) for tissue-clearing. Note: both products are required are for the clearing procedure.
$138.86 - $401.94
Specifications
| For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
|---|---|
| Product Line | CUBIC |
| For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
| Form | Liquid |
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|
T374125ML
|
TCI America™
T374125ML |
25 mL |
Each for $138.86
|
|
|||||
|
T3741100ML
|
TCI America™
T3741100ML |
100 mL |
Each for $401.94
|
|
|||||
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-R+(M) (T3741) is used for RI matching while CUBIC-L (T3740) is used for delipidation.
Note: Both T3741 and T3740 are required for the CUBIC clearing process.
Specifications
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |