missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCI America™ Tissue-Clearing Reagent CUBIC-P
CUBIC-P is a CUBIC reagent for mouse perfusion, which efficiently aids perfusion fixation.
$102.32 - $336.17
Specifications
| For Use With (Equipment) | Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device |
|---|---|
| Product Line | CUBIC |
| For Use With (Application) | Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications |
| Form | Liquid |
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|
T378225ML
|
TCI America™
T378225ML |
25 mL |
Each for $102.32
|
|
|||||
|
T3782100ML
|
TCI America™
T3782100ML |
100 mL |
Each for $336.17
|
|
|||||
Description
- The CUBIC technique enables cyclopedic imaging at a single-cell resolution following whole-body and whole-organ clearing.
- CUBIC-P is an optional addition to the CUBIC clearing protocol, and can be used to improve perfusion fixation.
Note: Both T3741 and T3740 are required for the CUBIC clearing process.
Specifications
| Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device | |
| CUBIC | |
| Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications | |
| Liquid |